Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Arlington Scientific Color Staph Test Kit
Rapid color agglutination test to detect clumping factor and/or protein-A in Staphylococcus aureus in cultures
$104.00 - $196.00
Specifications
Certifications/Compliance | CPT code 87449, 510(k)- K830783 |
---|---|
Detectable Analytes | MRSA |
Sensitivity | >99% |
Type | Color Staph Kit |
Includes | Liquid Controls |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
22-415-234
|
Arlington Scientific
285050 |
50 Tests |
Pack of 50 for $104.00
|
|
|||||
22-415-235
|
Arlington Scientific
285150 |
150 Tests |
Pack of 150 for $196.00
|
|
|||||
Description
- Color Staph latex reagent reacts with characteristics
- Positive reaction: aggregation of smooth latex suspension, red agglutination with blue background
- Detects MRSA
- Includes liquid controls
- Sensitivity: > 99%
- Specificity: > 99%
- Certifications:
- CPT code 87449
- 510(k)- K830783
Specifications
CPT code 87449, 510(k)- K830783 | |
>99% | |
Liquid Controls | |
10 to 30 sec. |
MRSA | |
Color Staph Kit | |
> 99% |
Safety and Handling
ShelfLife : Up to 24 Months
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title