Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Arlington Scientific Color Staph Test Kit
Rapid color agglutination test to detect clumping factor and/or protein-A in Staphylococcus aureus in cultures
Supplier: Arlington Scientific 285150
Description
- Color Staph latex reagent reacts with characteristics
- Positive reaction: aggregation of smooth latex suspension, red agglutination with blue background
- Detects MRSA
- Includes liquid controls
- Sensitivity: > 99%
- Specificity: > 99%
- Certifications:
- CPT code 87449
- 510(k)- K830783
Specifications
CPT code 87449, 510(k)- K830783 | |
>99% | |
Liquid Controls | |
> 99% |
MRSA | |
Color Staph Kit | |
150 Tests | |
10 to 30 sec. |
Safety and Handling
ShelfLife : Up to 24 Months
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction