Learn More
Remco™ Color-Coded Transport Storage Tub Lid
Remco™ Color-Coded Transport Storage Tub Lid is designed to help keep contaminants out of stored products.
Supplier: Remco™ 69125
Description
- Designed for use with Remco™ Transport Storage Tubs
- They are manufactured from high-density polyethylene
- Suitable for transporting, freezing, and storing
- This model has an angled edge that fits securely on angled tubs
- These lids meet all USDA specifications
- Commonly used for meat, seafood, poultry, commercial fishing, confectionery, bakery, and grain applications
Specifications
Tub Lid | |
Polyethylene | |
FDA Compliant | |
46.9 x 26.9 x 1.7 in. |
White | |
Transport Storage Tub | |
1 |