Learn More
Eisco™ Class A Glass Volumetric Flasks with Polypropylene Stopper
SCI_ED
Eisco™ Class A Glass Volumetric Flasks with Polypropylene Stopper features a single graduation mark and a fitted chemical resistant stopper.
$7.52 - $17.74
Specifications
Autoclavable Autoclavable
Bottom Shape Flat
Class A
Closure Color White
Closure Material Polypropylene
View More Specs
Products 3
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
S89443
SCI_ED
View Documents
Eisco™
CH0446GWT
250 mL
Each for $7.52
Add to cart
S89445
SCI_ED
View Documents
Eisco™
CH0446HWT
500 mL
Each for $8.81
Add to cart
S89446
SCI_ED
View Documents
Eisco™
CH0446IWT
1000 mL
Each for $17.74
Add to cart
Description
Description
  • Include chemical resistant polypropylene stopper
  • Flat bottom design
  • Single graduation mark
  • White printed markings
  • Class A, in compliance with ASTM-E288
  • Autoclavable
Warranty and Services
1 year limited manufacturer warranty
Specifications
Specifications
Autoclavable
A
Polypropylene
Clear
No
Yes
Long
Flat
White
Stopper
Borosilicate Glass 3.3
Prepare chemical solution
Beaded