Learn More
United Scientific™ Class A Gaduated Cylinder with Double Scale and Stopper, Individually Certified
Graduated Cylinders that are graduated 'To Deliver'.
$43.25 - $289.00
Specifications
Certifications/Compliance | ASTM E1272 |
---|---|
Class | A |
Type | Graduated Cylinder |
Material | Borosilicate Glass |
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
S00806
|
United Scientific™
CY298010 |
10 mL |
Each for $43.25
|
|
|||||
S00807
|
United Scientific™
CY298025 |
25 mL |
Each for $45.75
|
|
|||||
S00808
|
United Scientific™
CY298050 |
50 mL |
Each for $48.25
|
|
|||||
S00809
|
United Scientific™
CY2980100 |
100 mL |
Each for $54.00
|
|
|||||
S00810
|
United Scientific™
CY2980250 |
250 mL |
Each for $81.00
|
|
|||||
S00811
|
United Scientific™
CY2980500 |
500 mL |
Each for $113.00
|
|
|||||
S00812
|
United Scientific™
CY29801000 |
1000 mL |
Each for $209.00
|
|
|||||
S00813
|
United Scientific™
CY29802000 |
2000 mL |
Each for $289.00
|
|
|||||
Description
- Cylinders comply with ASTM E1272, Class A, meets USP standards, individually serialized and certified
- Each cylinder includes a hollow glass stopper
- Double metric scale, calibrated to contain
- Borosilicate glass cylinders feature heavy uniform wall tubing and strong, stable, hexagonal bases
- All cylinders come with bumper guards
Specifications
ASTM E1272 | |
A | |
Graduated Cylinder | |
Borosilicate Glass |