Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
United Scientific™ Class A Gaduated Cylinder with Double Scale and Stopper, Individually Certified
Graduated Cylinders that are graduated 'To Deliver'.
Supplier: United Scientific™ CY298050
This item is not returnable.
View return policy
Description
- Cylinders comply with ASTM E1272, Class A, meets USP standards, individually serialized and certified
- Each cylinder includes a hollow glass stopper
- Double metric scale, calibrated to contain
- Borosilicate glass cylinders feature heavy uniform wall tubing and strong, stable, hexagonal bases
- All cylinders come with bumper guards
Specifications
Graduated Cylinder | |
A | |
Borosilicate Glass | |
1 mL |
ASTM E1272 | |
0.25 mL | |
50 mL |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction