Learn More
MilliporeSigma™ Supelco™ ChromegaChiral™ Chiral CC3 HPLC Column, 5 μm Particle Size
ChromegaChiral™ CC3 (amylose tris(5-chloro-2-methyl- phenylcarbamate) is a new product for high resolution chiral separations based on a new halogenated carbohy- drate based chiral stationary phase.
$2556.00 - $23195.00
Specifications
Particle Size | 5 μm |
---|---|
Pore Size | 1000 Å |
Stationary Phase | Amylose, carbamate |
Product Line | ChromegaChiral |
Matrix | Amylose, carbamate Phase |
Catalog Number | Mfr. No. | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
11-100-5843
|
MilliporeSigma™ Supelco™
ES5680 |
2.1 mm | 10 cm |
Each for $4,437.00
|
|
||||
11-100-6344
|
MilliporeSigma™ Supelco™
ES5683 |
2.1 mm | 15 cm |
Each for $2,556.00
|
|
||||
11-100-5563
|
MilliporeSigma™ Supelco™
ES5687 |
2.1 mm | 25 cm |
Each for $20,415.00
|
|
||||
11-100-6342
|
MilliporeSigma™ Supelco™
ES5681 |
3 mm | 10 cm |
Each for $2,556.00
|
|
||||
11-100-6345
|
MilliporeSigma™ Supelco™
ES5684 |
3 mm | 15 cm |
Each for $2,556.00
|
|
||||
11-100-6159
|
MilliporeSigma™ Supelco™
ES5688 |
3 mm | 25 cm |
Each for $2,975.00
|
|
||||
11-100-6343
|
MilliporeSigma™ Supelco™
ES5682 |
4.6 mm | 10 cm |
Each for $2,556.00
|
|
||||
11-100-6346
|
MilliporeSigma™ Supelco™
ES5685 |
4.6 mm | 15 cm |
Each for $2,556.00
|
|
||||
11-100-6160
|
MilliporeSigma™ Supelco™
ES5689 |
4.6 mm | 25 cm |
Each for $2,975.00
|
|
||||
11-100-6161
|
MilliporeSigma™ Supelco™
ES5690 |
10 mm | 25 cm |
Each for $2,975.00
|
|
||||
11-100-6347
|
MilliporeSigma™ Supelco™
ES5686 |
20 mm | 15 cm |
Each for $2,556.00
|
|
||||
11-100-5703
|
MilliporeSigma™ Supelco™
ES5691 |
20 mm | 25 cm |
Each for $9,335.00
|
|
||||
11-100-5554
|
MilliporeSigma™ Supelco™
ES5692 |
30 mm | 25 cm |
Each for $23,195.00
|
|
||||
Specifications
5 μm | |
Amylose, carbamate | |
Amylose, carbamate Phase | |
Chiral HPLC |
1000 Å | |
ChromegaChiral | |
Chiral |
ChromegaChiral is a trademark of ES Industries