Learn More
Carolina™ Individual Bacterial Cultures (N-Z)
Bacteria cultures have wide applications in the food and chemical industries because of their highly diverse metabolic activities.
Specifications
For Use With (Application) | Ideal for food and chemical industries because of their highly diverse metabolic activities. |
---|---|
Storage Requirements | 20°C to 22°C |
Quantity | Ea. |
Includes | Carolina™ techniques for studying bacteria and fungi manual |
Sufficient For | Class of 30 students |
Alert:
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Catalog Number | Mfr. No. | Media Type | Packaging | Scientific Name | Temperature (Metric) Range | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Media Type | Packaging | Scientific Name | Temperature (Metric) Range | Price | Quantity & Availability | ||
S07410ND
|
Carolina Biological Supply
155255A |
Nutrient Agar | Vial | Pseudomonas fluorescens | 25°C | N/A | N/A | ||
This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-955-1177. Provide the NEED BY DATE to the customer service agent. | |||||||||
S07200ND
|
Carolina Biological Supply
155420 |
Nutrient Agar | Tube | Sarcina lutea | 25°C |
Each for $14.75
|
N/A | ||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
S07203ND
|
Carolina Biological Supply
155554A |
Brain Heart Infusion Agar | Vial | Staphylococcus aureus (Coagulase positive) | 37°C | N/A | N/A | ||
This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-955-1177. Provide the NEED BY DATE to the customer service agent. | |||||||||
S07196ND
|
Carolina Biological Supply
155240A |
Nutrient Agar | Vial | Proteus vulgaris | 37°C | N/A | N/A | ||
This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-955-1177. Provide the NEED BY DATE to the customer service agent. | |||||||||
S07412ND
|
Carolina Biological Supply
155640A |
Tryptic Soy Agar with 5% Sheep Blood or Dextrose Starch Agar | Vial | Streptococcus salivarius | 37°C | N/A | N/A | ||
This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-955-1177. Provide the NEED BY DATE to the customer service agent. | |||||||||
S07195ND
|
Carolina Biological Supply
155250A |
Nutrient Agar | Vial | Pseudomonas aeruginosa | 37°C | N/A | N/A | ||
This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-955-1177. Provide the NEED BY DATE to the customer service agent. | |||||||||
S07202ND
|
Carolina Biological Supply
155452 |
Nutrient Agar | Tube | Serratia marcescens D1 | 25°C |
Each for $14.75
|
N/A | ||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
S07414ND
|
Carolina Biological Supply
155351A |
Nutrient Agar | Vial | Salmonella typhimurium | 37°C | N/A | N/A | ||
This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-955-1177. Provide the NEED BY DATE to the customer service agent. | |||||||||
S07197ND
|
Carolina Biological Supply
155255 |
Nutrient Agar | Tube | Pseudomonas fluorescens | 25°C |
Each for $14.75
|
N/A | ||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
S07409ND
|
Carolina Biological Supply
155239A |
Nutrient Agar | Vial | Proteus mirabilis | 37°C | N/A | N/A | ||
This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-955-1177. Provide the NEED BY DATE to the customer service agent. | |||||||||
S07211ND
|
Carolina Biological Supply
155722 |
Photobacterium Agar | Tube | Vibrio fischeri | 25°C |
Each for $14.75
|
N/A | ||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
S07411ND
|
Carolina Biological Supply
155470A |
Nutrient Agar | Vial | Shigella flexneri | 37°C |
Each for $23.25
|
N/A | ||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
S07405ND
|
Carolina Biological Supply
155556A |
Nutrient Agar | Vial | Staphylococcus epidermidis | 37°C | N/A | N/A | ||
This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-955-1177. Provide the NEED BY DATE to the customer service agent. | |||||||||
S07201ND
|
Carolina Biological Supply
155450 |
Nutrient Agar | Tube | Serratia marcescens | 25°C |
Each for $14.75
|
N/A | ||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
S07426ND
|
Carolina Biological Supply
155450A |
Nutrient Agar | Vial | Serratia marcescens | 25°C | N/A | N/A | ||
This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-955-1177. Provide the NEED BY DATE to the customer service agent. | |||||||||
S07401ND
|
Carolina Biological Supply
155620A |
Tryptic Soy Agar with 5% Sheep Blood | Vial | Streptococcus pneumoniae | 37°C | N/A | N/A | ||
This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-955-1177. Provide the NEED BY DATE to the customer service agent. | |||||||||
S07400ND
|
Carolina Biological Supply
155630A |
Tryptic Soy Agar with 5% Sheep Blood | Vial | Streptococcus pyogenes | 37°C | N/A | N/A | ||
This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-955-1177. Provide the NEED BY DATE to the customer service agent. | |||||||||
S07429ND
|
Carolina Biological Supply
155556 |
Nutrient Agar | Tube | Staphylococcus epidermidis | 37°C | N/A | N/A | ||
This item is living or perishable, a NEED BY DATE is REQUIRED for proper delivery. Orders must be placed through Customer Service 800-955-1177. Provide the NEED BY DATE to the customer service agent. | |||||||||
Description
Bacteria are involved in many of the events that make life possible for other organisms, including humans. Fossils of prokaryotes similar to today's bacteria have been found in 3.5 billion-year-old rocks. A mastery of bacteria is the key to understanding life on earth in the past, present, and future.
- Labeled by genus and species name and media on which they are cultured
- All Gram staining based on 24 hr. cultures
- Pure and high-quality
- For every level of study
- Easy-to-use media
Specifications
Ideal for food and chemical industries because of their highly diverse metabolic activities. | |
Ea. | |
Class of 30 students |
20°C to 22°C | |
Carolina™ techniques for studying bacteria and fungi manual |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.