Learn More
Carolina™ Individual Bacterial Cultures (N-Z)
Bacteria cultures have wide applications in the food and chemical industries because of their highly diverse metabolic activities.
Supplier: Carolina Biological Supply 155450
Alert:
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000
Description
Bacteria are involved in many of the events that make life possible for other organisms, including humans. Fossils of prokaryotes similar to today's bacteria have been found in 3.5 billion-year-old rocks. A mastery of bacteria is the key to understanding life on earth in the past, present, and future.
- Labeled by genus and species name and media on which they are cultured
- All Gram staining based on 24 hr. cultures
- Pure and high-quality
- For every level of study
- Easy-to-use media
Specifications
Ideal for food and chemical industries because of their highly diverse metabolic activities. | |
20°C to 22°C | |
Serratia marcescens | |
Class of 30 students | |
Bacteria Culture |
Tube | |
Ea. | |
Carolina™ techniques for studying bacteria and fungi manual | |
Nutrient Agar | |
25°C |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.