Learn More
Thermo Scientific Chemicals Carbonate, 0.5M buffer soln., pH 9.6
$102.00 - $164.00
Specifications
Concentration | 0.5 M buffer soln. |
---|---|
pH Range | 9.6 |
Type | Carbonate buffer soln. |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AAJ62610AK
|
Thermo Scientific Chemicals
J62610AK |
250 mL |
Each for $102.00
Order in Bulk
Request bulk or custom formats |
|
|||||
AAJ62610AP
|
Thermo Scientific Chemicals
J62610AP |
500 mL |
Each for $164.00
Order in Bulk
Request bulk or custom formats |
|
|||||
Description
Sodium bicarbonate buffer system is important for several different cellular processes. It is one of the major buffering systems used to maintain the pH of mammalian blood and is also important in the acid-base homeostasis of living things, including humans. It tends to maintain a relatively constant plasma pH and counteract any force that would alter it. It is used to neutralize the pH of the chyme leaving the stomach and entering the small intestine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSodium bicarbonate buffer system is important for several different cellular processes. It is one of the major buffering systems used to maintain the pH of mammalian blood and is also important in the acid-base homeostasis of living things, including humans. It tends to maintain a relatively constant plasma pH and counteract any force that would alter it. It is used to neutralize the pH of the chyme leaving the stomach and entering the small intestine.
Solubility
It is soluble in water.
Notes
Store at 4°C in tightly closed container. Avoid inhalation. Wash thoroughly after handling. Avoid prolonged or repeated exposure.
Specifications
0.5 M buffer soln. | |
9.6 | |
Carbonate buffer soln. |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.