Learn More
MilliporeSigma™ Calbiochem™ ProteoExtract™ Cytosol/Mitochondria Fractionation Kit
Used for isolation of enriched mitochondrial fraction from cytosol.
Supplier: MilliporeSigma™ QIA881KIT
Description
- Designed for extraction of mammalian cells to yield enriched mitochondrial cytosolic fractions
- Downstream analyses, such as Western blotting
- Useful for isolation of highly enriched mitochondrial fraction from cytosol
- Packaging: Fibre Case
Specifications
100 Extractions/Kit | |
Mitochondria Extraction Buffer, Cytosol Extraction Buffer, Protease Inhibitor Cocktail, Reducing Reagent, Cytochrome c Antibody, and user protocol. | |
2°C to 8°C |