Learn More
Bullard™ 41 Series CO Monitor and Filter
SDP
Provides 7-stage filtration combined with carbon monoxide (CO) monitoring.
Supplier:  Bullard™ COHP41P2
 View more versions of this product
Catalog No. 19-100-600
Add to Cart
Description
Description
Monitors and filters may also be used to supply drier, cleaner air to tools and pneumatic systems to reduce downtime and repair costs
7-Stage Filtration
  • Water is removed by condensation of air in outer cylinder
  • Carded cotton removes particulates
  • Activated alumina absorbs oil and moisture
  • Activated charcoal removes odors and moisture
  • Felt material removes particulates
  • Carded cotton removes particulates
  • Respiratory felt at final stage acts as a final filter before air is transferred to worker or air driven tools
Design
  • Steel construction for rugged industrial environments
  • Large disposable filter cartridge for long life
  • Carrying handle available for easy transport
  • Weighs only 23 pounds
  • Two or six outlets
  • Free standing or wall mountable
Warranty and Services
One-year limited warranty
Specifications
Specifications
2 outlet
41 Series Airline Respirator
7-stage filtration filter unit, CO monitor with auto calibration, audible and visual alarms for high co and low battery and 6 ft. 110 V power cord, pressure gauge, 125 psig relief valve and pressure regulator
23.5 in.
22 lb.
CO Monitor and Filter Kit
75 cfm
Steel
COHP41P2
60 cm
10 kg