Learn More
United Scientific™ Borosilicate Glass Narrow Mouth Erlenmeyer Flasks
Borosilicate glass narrow mouth Erlenmeyer flasks are designed to give the best combination of thermal shock resistance and physical strength.
$25.75 - $87.00
Specifications
Material | Borosilicate Glass (3.3) |
---|---|
For Use With (Application) | General Lab Use |
Autoclavable | Autoclavable |
Mouth Style | Narrow |
Sterility | Non-sterile |
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
S111085
|
United Scientific™
FG498010 |
10 mL |
Pack of 12 for $38.25
|
|
|||||
S111086
|
United Scientific™
FG498025 |
25 mL |
Pack of 12 for $41.00
|
|
|||||
S111087
|
United Scientific™
FG498050 |
50 mL |
Pack of 12 for $39.75
|
|
|||||
S111088
|
United Scientific™
FG4980100 |
100 mL |
Pack of 12 for $39.75
|
|
|||||
S111089
|
United Scientific™
FG4980125 |
125 mL |
Pack of 12 for $51.00
|
|
|||||
S111090
|
United Scientific™
FG4980150 |
150 mL |
Pack of 6 for $25.75
|
|
|||||
S111091
|
United Scientific™
FG4980250 |
250 mL |
Pack of 12 for $63.50
|
|
|||||
S111092
|
United Scientific™
FG4980500 |
500 mL |
Pack of 6 for $63.50
|
|
|||||
S111093
|
United Scientific™
FG49801000 |
1000 mL |
Pack of 6 for $87.00
|
|
|||||
S111094
|
United Scientific™
FG49802000 |
2000 mL |
Each for $27.00
|
|
|||||
S111095
|
United Scientific™
FG49803000 |
3000 mL |
Each for $47.25
|
|
|||||
S111096
|
United Scientific™
FG49805000 |
5000 mL |
Each for $80.00
|
|
|||||
Description
Borosilicate glass flasks are designed to give the best combinations of thermal shock resistance and physical strength.- Narrow Mouth, Erlenmeyer
- Feature heavy-duty rims.
- Graduations in durable white enamel.
Specifications
Borosilicate Glass (3.3) | |
Autoclavable | |
Non-sterile |
General Lab Use | |
Narrow | |
Flat |