Learn More
United Scientific™ Borosilicate Glass Distilling Flask
Distilling Flasks ideal for distillation applications.
Supplier: United Scientific™ FG4620125
This item is not returnable.
View return policy
Description
- Distilling flasks are made from low-expansion borosilicate glass
- Uniform wall thickness provides superior mechanical strength and shock resistance
- Long side arm is ideal for distillation applications
- Feature large white block letters for easy identification of capacity
Specifications
125 mL | |
Long Side Arm | |
Autoclavable | |
No | |
Non-sterile |
Borosilicate Glass | |
12 | |
Reusable | |
Rimmed | |
Round |