Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TerraCycle™ Biodegradable Plastic Zero Waste Box™
![SDP](/content/dam/fishersci/glyphs/sdp.png)
![SDP](/content/dam/fishersci/glyphs/sdp.png)
![SCI_ED](/content/dam/fishersci/glyphs/scied.png)
Recycle Biodegradable Plastic using TerraCycle's Zero Waste Box system. Each box includes signage, a built-in liner, shipping to and from your location, and processing of the materials.
Supplier: TerraCycle™ CS161S
Description
WHAT YOU CAN RECYCLE
Please send any brand and type of biodegradable or compostable plastic good. This includes PLA cups, knives, forks, spoons, bowls, and plates.
DO NOT SEND in organic matter, non-biodegradable plastic, or paper or wood compostables. Small amounts of residual food or beverage waste is acceptable.
Specifications
39.66 L | |
11 x 11 x 20 in. | |
Not Autoclavable | |
Recycling Box |
Brown, Green, White | |
Small | |
Cardboard |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction