Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dynalon™ Bio-bin™ Waste Disposal Containers
Bio-bin™ is the ideal solution for biohazard or non-hazardous waste disposal. Reduce cost, weight, transport, CO2 emissions, storage space!
$53.25 - $324.00
Catalog Number | Mfr. No. | Capacity (Metric) | Description | Dimensions (L x W x H) | For Use With (Application) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Description | Dimensions (L x W x H) | For Use With (Application) | Price | Quantity & Availability | ||
S29243
|
Dynalon™
7973030006/EMD |
6 L | Bio-Bin™ Loop Model 6L | 6 x 6 x 15 in. | Biohazard or non-hazardous waste |
Case of 40 for $324.00
|
|
||
S29244
|
Dynalon™
7973030030/EMD |
30 L | Bio-Bin™ Loop Model 30L | 11 x 11 x 16.3 in. | Biohazard or non-hazardous waste |
Case of 10 for $241.00
|
|
||
S29245
|
Dynalon™
7973036100/EMD |
6 L | Bio-Bin™ U Stand | 7.1 x 10.8 x 12.32 in. | Holding 6L Bio-Bin™ at a 30 degree angle |
Each for $53.25
|
|
||
S29246
|
Dynalon™
7973036200/EMD |
6 L | Bio-Bin™ Z Stand | 6.7 x 6.7 x 7.87 in. | Holding 6L Bio-Bin™ upright |
Each for $53.25
|
|
||
S29247
|
Dynalon™
7973036300/EMD |
30L | Bio-Bin™ M Stand | 16.85 x 7.12 x 11.25 in. | Holding 30L Bio-Bin™ |
Each for $164.00
|
|
||
S29242
|
Dynalon™
7973030002/EMD |
2.5 L | Bio-Bin™ Loop Model 2.5L | 9 x 4.3 x 4.7 in. | Biohazard or non-hazardous waste | Item Discontinued This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | ||
Description
- Space efficient, lightweight fold-flat design
- Built-in lid and handles
- Temporary and permanent seals
- Stackable post use
- UN 3291 Approved
- Printed user instructions and user ID/date information on lid
- Waterproof construction
Specifications
UN 3291 | |
Autoclavable |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title