Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dynalon™ Bio-bin™ Waste Disposal Containers
Bio-bin™ is the ideal solution for biohazard or non-hazardous waste disposal. Reduce cost, weight, transport, CO2 emissions, storage space!
Supplier: Dynalon™ 7973036200/EMD
Description
- Space efficient, lightweight fold-flat design
- Built-in lid and handles
- Temporary and permanent seals
- Stackable post use
- UN 3291 Approved
- Printed user instructions and user ID/date information on lid
- Waterproof construction
Specifications
Stand | |
6 L | |
Bio-Bin™ Z Stand | |
199.9 mm | |
Z-shaped | |
Autoclavable | |
6L Bio-Bin™ | |
6.7 in. |
Stainless Steel | |
UN 3291 | |
6.7 x 6.7 x 7.87 in. | |
170.18 mm | |
170.18 mm | |
Holding 6L Bio-Bin™ upright | |
6.7 in. | |
7.87 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction