Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisher Science Education™ Benedict's Solution, Qualitative
$7.40 - $45.93
Specifications
Chemical Name or Material | Benedict's Solution, Qualitative |
---|---|
Color | Blue |
CAS | 63126-89-6, 6132-04-3, 6132-4-3, 7732-18-5 |
Assay Percent Range | <2% w/v |
Density | 1.2g/mL |
Specifications
Benedict's Solution, Qualitative | |
63126-89-6, 6132-04-3, 6132-4-3, 7732-18-5 | |
1.2g/mL | |
Liquid |
Blue | |
<2% w/v | |
Solution |
Safety and Handling
ShelfLife : 2 years
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title