Learn More
Fisher Science Education™ Benedict's Solution, Qualitative
SCI_ED
 View more versions of this product
Catalog No. S25193
Edge
Add to Cart
Description
Description
Specifications
Specifications
Benedict's Solution, Qualitative
63126-89-6, 6132-04-3, 6132-4-3, 7732-18-5
1.2g/mL
500 mL
Blue
<2% w/v
Solution
Liquid
Product Suggestions
Product Suggestions
Safety and Handling
Safety and Handling
ShelfLife : 2 years