Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Port-A-Cul™ Anaerobic Collection and Transport Systems

Versatile—use for fluid, tissue, or swab specimens

Supplier:  BD 221609

 View more versions of this product

Catalog No. B21609

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View alternate product


Description

Description

  • Collect and maintain specimens at 20° to 25°C for up to 72 hours
  • Ensure aerobic, anaerobic, microaerophilic or facultative microorganism viability during transport from patient to laboratory (including Campylobacter jejuni)
  • Reduced medium formulated to protect oxygen-sensitive microbes
  • Media contain an indicator that continuously monitors the reduced media status
  • Collect specimen on a swab, as a fluid, or as a tissue specimen
  • Available as tube system (capacity 11mL), jar system (capacity 20mL), or vial system (capacity 5mL)
  • Sterile Package contains either one tube and two polyester-tipped wooden swabs, one vial or one jar
  • Screw-cap jar is 3 in. tall and has a 1 in.-wide mouth; not for use with fluids
  • Fluid Collection Kit contains one sterile-packaged vial containing media and a 5cc syringe with a 22-gauge needle and Luer-Lok™ closure

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Port-A-Cul™ Vial
Used for transport of fluid specimens
20°C to 25°C
Sterile
10 Pk.
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit