Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD Difco™ Vibrio cholerae Antisera
Quick reactive Gold standard Vibrio cholerae Antisera
$417.00
Specifications
Certifications/Compliance | CE Marked |
---|---|
Content And Storage | Refrigerate, store at 2°C to 8°C |
Product Type | Microbiology Media |
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
D243047
|
BD
224301 |
Microbiology Media |
Each for $417.00
|
|
|||||
DF2431-47-0
|
BD
224311 |
Microbiology Media |
Each for $413.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
DF2432-47-9
|
BD
224321 |
Microbiology Media |
Each for $403.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
- Product type: Antigens and Antisera
- Groupings: U through Z Products
- Sub type: Vibrio cholerae
- Type: Antisera
- Lyophilized, rehydrate to 3mL
- Sufficient reagent for 20 slide tests
- Available with 3 different antiserum types and O Antibodies
- Shelf Life: 1095 days
Rabbit serum for slide agglutination test
Certifications
CE marked
Specifications
CE Marked | |
Refrigerate, store at 2°C to 8°C | |
Microbiology Media |
Safety and Handling
ShelfLife : 1095 Days
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title