Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD Difco™ Vibrio cholerae Antisera
Quick reactive Gold standard Vibrio cholerae Antisera
Supplier: BD 224301
This item is not returnable.
View return policy
Description
- Product type: Antigens and Antisera
- Groupings: U through Z Products
- Sub type: Vibrio cholerae
- Type: Antisera
- Lyophilized, rehydrate to 3mL
- Sufficient reagent for 20 slide tests
- Available with 3 different antiserum types and O Antibodies
- Shelf Life: 1095 days
Rabbit serum for slide agglutination test
Certifications
CE marked
Specifications
CE Marked | |
Refrigerate, store at 2°C to 8°C | |
Microbiology Media |
Safety and Handling
ShelfLife : 1095 Days
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction