Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azer Scientific Formagel™ Tissue Fixation and Transport Gel
SDP

Solution to leaking formalin problems for improved histology studies

Supplier:  Azer Scientific PFG60

 View more versions of this product

Catalog No. 22-026-357

  • $11.55 / Case of 50

    Save 87%

    Reg : $91.90

    The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
    Learn More
Add to cart

Description

Description

  • Maintains specimen integrity
  • Fixation results identical to standard 10% Neutral Buffered Formalin
  • Viscoscity enables safe transport without spills

For leak-free transport and fixation of tissue specimens. Compatible with routine, special and IHC staining procedures.

Specifications

Specifications

Screw-on
10% Neutral Buffered Formalin
30mL
Polypropylene
Clear
Formagel™ Prefilled Container
Routine, special, IHC staining procedures
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit