Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SCS Anti-Fatigue Vinyl Mat Kit
A suitable flooring component in personnel grounding Footwear/Flooring System
Specifications
Color | Gray |
---|---|
Length (English) | 36 in. |
Length (Metric) | 91.44 cm |
Material | Vinyl |
Thickness (English) | 0.350 in. |
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
18-688-344
|
SCS
AFM3660L1VGRF |
36 x 60 in. | Item Discontinued This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
18-688-345
|
SCS
AFM36720L1VGR |
50 ft. x 36 in. | Item Discontinued This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
Description
- Reversible Dissipative Rubber Floor Matting
Provides a flooring surface that does not generate a static charge and will act as part of the path to ground for an operator wearing ESD footwear - Rtt: < 1 x 108 to < 1 x 1011 ohms per ANSI/ESD 7.1 and ESD TR53Use when meeting ANSI/ESD S20.20 flooring required limit is not required
- Low Profile
Reduces trip hazards and designed for use in high traffic areas and good for use under chairs and carts. - Made in the United States of America
This item is stocked in Sanford, NC
Specifications
Gray | |
91.44 cm | |
0.350 in. |
36 in. | |
Vinyl |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title