Learn More
SCS Anti-Fatigue Vinyl Mat Kit
A suitable flooring component in personnel grounding Footwear/Flooring System
Supplier: SCS AFM3660L1VGRF
Description
- Reversible Dissipative Rubber Floor Matting
Provides a flooring surface that does not generate a static charge and will act as part of the path to ground for an operator wearing ESD footwear - Rtt: < 1 x 108 to < 1 x 1011 ohms per ANSI/ESD 7.1 and ESD TR53Use when meeting ANSI/ESD S20.20 flooring required limit is not required
- Low Profile
Reduces trip hazards and designed for use in high traffic areas and good for use under chairs and carts. - Made in the United States of America
This item is stocked in Sanford, NC
Specifications
36 x 60 in. | |
36 in. | |
91.44 cm | |
152.4 cm |
0.350 in. | |
Gray | |
Vinyl | |
60 in. |