Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Anode Carbon Composite Toe

Men’s 4.5 in. 100% Non-Metallic ESD Carbon Composite Toe 4x4 Hiker

Supplier:  Carolina™ CA55261002E

 View more versions of this product

Catalog No. 19-154-134

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Description

Description

  • Brown
  • Bandit Potting Soil Leather
  • Composite Safety Toe
  • ESD
  • 6 Inch
  • Broad Toe
Specifications

Specifications

Brown
10
Composite
Leather
Rubber
2E
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit