Learn More
Thermo Scientific Chemicals Ammonium, Quant|r Test Strips
Supplier: Thermo Scientific Chemicals 023267QA
Alert:
H314; Causes severe skin burns and eye damage. P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a; Do not breathe dust/fume/gas/mist/vapours/spray. IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations.
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Fully miscible
- Liquid form
Specifications
Liquid | |
Reagents and strips to measure 10-400ppm NH4+1 in aqueous solution | |
Ambient | |
Fully Miscible |