Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abbott™ Impact RPR™ Test Kits

IMPACT RPR™ Card Test is a non-treponemal test for the rapid detection and quantitative determination of syphilis in serum or plasma.

Supplier:  Abbott™ 60C3

 View more versions of this product

Catalog No. WA-60C3


Edge

Due to product restrictions, please to purchase or view availability for this product.

Explore available promotions

Alert:

RPR Card Antigen Suspension contains sodium azide, which is harmful if swallowed. Contact with acids liberates very toxic gas. After contact with skin, wash immediately with plenty of water. Sodium azide may react with lead and copper plumbing to form highly explosive metal azides. On disposal, flush with a large volume of water to prevent azide buildup.


Includes: RPR Test Kits: Antigen suspension, dispensing bottle with needle and sheath, pipet/stirrers, and plastic test cards with ten or thirty 18mm diameter wells that will not warp or bow
This item is not returnable. View return policy

Description

Description

  • Classified as a standard test for syphilis by the Centers for Disease Control and Prevention (CDC)
  • Non-treponemal test for the serological detection of syphilis
  • Assay utilizes carbon particle technology

Order Info

Controls available separately in both dry and liquid form.

Specifications

Specifications

Reagin (Syphilis)
RPR Test Kit
Antigen suspension, dispensing bottle with needle and sheath, pipet/stirrers, Plastic test cards with ten 18mm diameter wells
500 Tests
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.