Learn More
Abbott™ Impact RPR™ Test Kits
IMPACT RPR™ Card Test is a non-treponemal test for the rapid detection and quantitative determination of syphilis in serum or plasma.
$423.00
Specifications
Detectable Analytes | Reagin (Syphilis) |
---|---|
Type | RPR Test Kit |
Includes | Antigen suspension, dispensing bottle with needle and sheath, pipet/stirrers, Plastic test cards with ten 18mm diameter wells |
Alert:
RPR Card Antigen Suspension contains sodium azide, which is harmful if swallowed. Contact with acids liberates very toxic gas. After contact with skin, wash immediately with plenty of water. Sodium azide may react with lead and copper plumbing to form highly explosive metal azides. On disposal, flush with a large volume of water to prevent azide buildup.
Includes: RPR Test Kits: Antigen suspension, dispensing bottle with needle and sheath, pipet/stirrers, and plastic test cards with ten or thirty 18mm diameter wells that will not warp or bow
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
WA-60C1
|
Abbott™
60C1 |
50 Tests |
Each for $68.75
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
WA-60C2
|
Abbott™
60C2 |
150 Tests |
Each for $146.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
WA-60C3
|
Abbott™
60C3 |
500 Tests |
Each for $423.00
|
|
|||||
Description
- Classified as a standard test for syphilis by the Centers for Disease Control and Prevention (CDC)
- Non-treponemal test for the serological detection of syphilis
- Assay utilizes carbon particle technology
Order Info
Controls available separately in both dry and liquid form.
Specifications
Reagin (Syphilis) | |
RPR Test Kit | |
Antigen suspension, dispensing bottle with needle and sheath, pipet/stirrers, Plastic test cards with ten 18mm diameter wells |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.