Learn More
MilliporeSigma™ Supelco™ Aldrich™ Rectangular TLC Developing Tanks, Lid Only
Aldrich™ rectangular TLC developing tanks, lid only may be used to develop TLC plates.
$38.85 - $110.30
Specifications
For Use With (Application) TLC
Product Line Aldrich
View More Specs
Products 6
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
11-101-1631
View Documents
MilliporeSigma™ Supelco™
Z4120661EA
Z204153
Each for $110.30
Add to Cart
11-101-1899
View Documents
MilliporeSigma™ Supelco™
Z4120741EA
Z204188
Each for $39.15
Add to Cart
11-101-1908
View Documents
MilliporeSigma™ Supelco™
Z4120821EA
Z204196
Each for $38.85
Add to Cart
11-101-1909
View Documents
MilliporeSigma™ Supelco™
Z4120901EA
Z204226
Each for $38.85
Add to Cart
11-101-1891
View Documents
MilliporeSigma™ Supelco™
Z1462181EA
Z126195 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
11-101-1921
View Documents
MilliporeSigma™ Supelco™
Z1462341EA
Z146226 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
TLC developing tanks are used to run TLC plates for separation. It is important to maintain saturated conditions during the analysis which can be achieved by using small tanks, filter paper liners and sufficient solvents. The tank can be equilibrated for 30 mins prior to running the plate to yield good results. Lid is used to cover the tank.
Specifications
Specifications
TLC
Aldrich
SDS
Documents
Documents
Product Certifications
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC