Learn More
MilliporeSigma™ Supelco™ Aldrich™ Rectangular TLC Developing Tanks, Lid Only
Aldrich™ rectangular TLC developing tanks, lid only may be used to develop TLC plates.
Supplier: MilliporeSigma™ Supelco™ Z1462181EA
Description
TLC developing tanks are used to run TLC plates for separation. It is important to maintain saturated conditions during the analysis which can be achieved by using small tanks, filter paper liners and sufficient solvents. The tank can be equilibrated for 30 mins prior to running the plate to yield good results. Lid is used to cover the tank.Specifications
TLC | |
Z126195 | |
Aldrich |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC