Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ accumet™ AE150 Benchtop pH Meter
Provides the essential functions needed for routine pH measurements without complicated features and processes at a budget-friendly price
Specifications
Accuracy (mV) | ±0.2 (±199.9), ±2 (Beyond) |
---|---|
Outputs | Recorder (±2000 mV) |
Range (pH) | 0 to 14 |
Range (mV) | ±1999 |
Resolution (mV) | 0.1 mV (±199.9 mV); 1 mV beyond ±200 mV |
Catalog Number | Includes | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Includes | Price | Quantity & Availability | ||||||
S30245
|
Meter, Electrode Stand, Literature, Universal Power Adapter (90-240V, 50/60 Hz) | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||||
S30244
|
Education set with probe and stand | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||||
S30246
|
Double Junction pH Electrode, Temperature Probe and Stand | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||||
S30247
|
3-in-1 set: 13-620-AE6 3-in-1 single junction gel pH/ATC electrode and stand | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||||
Description
- One to five point pH calibration
- Automatic buffer recognition with two pH buffer sets
- pH slope and offset display
- One point mV calibration
- One point ATC temperature probe calibration
- Automatic or manual temperature compensation
- Ready icon to indicate measurement stability
- Electrode condition icon to indicate status
- 100 data set storage with non-volatile memory
Specifications
±0.2 (±199.9), ±2 (Beyond) | |
0 to 14 | |
0.1 mV (±199.9 mV); 1 mV beyond ±200 mV | |
0.1°C | |
100 data sets | |
AC/DC 9 V adapter (110/220 VAC, 50-60 Hz) |
Recorder (±2000 mV) | |
±1999 | |
0.0°C to 100.0°C | |
Offset in 0.1 °C increments; Offset range: ±5 °C | |
BNC, 2.5 mm phono plug, reference pin tip, ground |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title