Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ accumet™ AE150 Benchtop pH Meter
SCI_ED

Provides the essential functions needed for routine pH measurements without complicated features and processes at a budget-friendly price

 View more versions of this product

Catalog No. S30247

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.


Description

Description

  • One to five point pH calibration
  • Automatic buffer recognition with two pH buffer sets
  • pH slope and offset display
  • One point mV calibration
  • One point ATC temperature probe calibration
  • Automatic or manual temperature compensation
  • Ready icon to indicate measurement stability
  • Electrode condition icon to indicate status
  • 100 data set storage with non-volatile memory
TRUSTED_SUSTAINABILITY
Specifications

Specifications

±0.01
±0.2 (±199.9), ±2 (Beyond)
3-in-1 set: 13-620-AE6 3-in-1 single junction gel pH/ATC electrode and stand
0.01
0.1 mV (±199.9 mV); 1 mV beyond ±200 mV
0.0°C to 100.0°C
Offset in 0.1 °C increments; Offset range: ±5 °C
BNC, 2.5 mm phono plug, reference pin tip, ground
Automatic/Manual (0°C to 100°C)
±0.3°C
Recorder (±2000 mV)
0 to 14
±1999
1 to 5 with automatic buffer recognition (ABR)
0.1°C
100 data sets
3 Year Meter Warranty; 6 Month Electrode Warranty
AC/DC 9 V adapter (110/220 VAC, 50-60 Hz)
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit