Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Axygen™ Accessories for Horizontal Gel Box, 15 cm
For use with 15 cm horizontal gel box
Supplier: Axygen™ HGB153515/EMD
Description
Specifications
Axygen Horizontal Gel Box, 15 cm | |
35-well Comb, 1.5 mm thickness | |
35 | |
1.5 mm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction