Learn More
Agilent BioTek 405LS Microplate Washer, IVD PROMO
The 405 LS washer has a well earned reputation as an economical plate washer that offers excellent performance and reliability for in vitro diagnostic use.
Specifications
Certifications/Compliance | CE and TUV Marked, RoHS Compliant |
---|---|
Bottles | 4 or 10 L (optional) |
Precision | <3% CV: 300 μL/well (96-well washing), <4% CV: 80 μL/well (384-well washing) |
Dispense Volume | 25 to 3000 μL/well |
Plate Type | 96-Well Plates, 384-Well Plates |
Catalog Number | Mfr. No. | Model | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Model | Includes | Price | Quantity & Availability | ||||
40-5LS-RSSIPM
|
Agilent BioTek
405LSRSSIPM |
405LSRS | Ultrasonic Advantage, 96-well Microplate Washing | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
40-5LU-VSSIPM
|
Agilent BioTek
405LUVSSIPM |
405LSUVS | Automated Internal 4-Buffer Switching, Ultrasonic Advantage, 96-/384-well Microplate Washing | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
- For In Vitro Diagnostic Use
- Washing protocols range from gentle cell washing to vigorous ELISA workflows
- Simple user interface for creation and execution of wash, dispense, aspirate, and maintenance protocols
- Extensive onboard software
For use under computer control, the Agilent BioTek™ LHC software used with the 405 LS is available in standard and 21 CFR Part 11 compliant versions. The optional Agilent BioTek™ BioStack is available for walk-away automation of up to 50 microplates.
Specifications
CE and TUV Marked, RoHS Compliant | |
<3% CV: 300 μL/well (96-well washing), <4% CV: 80 μL/well (384-well washing) | |
96-Well Plates, 384-Well Plates | |
Programmable in minutes and seconds, up to 60 min. | |
13.5 to 16.5 kg | |
High to Low Flow | |
50/60 Hz | |
14 in. | |
10 in. | |
17 in. | |
30 to 36 lb. |
4 or 10 L (optional) | |
25 to 3000 μL/well | |
<2 μL/well (96- & 384-well Plates); 96-tube Manifold For 96 Wells; 192-tube For 384 Wells | |
Microplate Washer | |
1 to 250 | |
10 to 85% | |
1 μL | |
35.6 cm | |
25.4 cm | |
43.2 cm | |
100/240 V |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.