Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
This item is not returnable.
View return policy
Description
- For In Vitro Diagnostic Use
- Washing protocols range from gentle cell washing to vigorous ELISA workflows
- Simple user interface for creation and execution of wash, dispense, aspirate, and maintenance protocols
- Extensive onboard software
For use under computer control, the Agilent BioTek™ LHC software used with the 405 LS is available in standard and 21 CFR Part 11 compliant versions. The optional Agilent BioTek™ BioStack is available for walk-away automation of up to 50 microplates.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
CE and TUV Marked, RoHS Compliant | |
<3% CV: 300 μL/well (96-well washing), <4% CV: 80 μL/well (384-well washing) | |
96-well Plate, 384-well Plate | |
<30 s | |
Microplate Washer | |
1 to 250 | |
10 to 85% | |
405LSRS | |
Ultrasonic Advantage, 96-well Microplate Washing | |
35.6 cm | |
25.4 cm | |
43.2 cm | |
100/240 V |
Optional (4 or 10 L) | |
25 to 3000 μL/well | |
<2 μL/well (96- & 384-well Plates); 96-tube Manifold For 96 Wells; 192-tube For 384 Wells | |
Programmable in minutes and seconds, up to 60 min. | |
13.5 to 16.5 kg | |
High to Low Flow | |
50/60 Hz | |
1 μL | |
14 in. | |
10 in. | |
17 in. | |
30 to 36 lb. |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction