Assay Details
Mature miRNA Sequence: | UCUCAGCUGCUGCCCUCUCCAG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-6787-3p | MIMAT0027475 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-6787-3p | ||
---|---|---|---|
miRBase Accession Number: | MI0022632 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.17: 82236668 - 82236728 [+] on Build GRCh38 | ||
Mature miRNA Sequence: | UCUCAGCUGCUGCCCUCUCCAG | ||
Species: | Human | ||
Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Mature miRNA Sequence: | UCUCAGCUGCUGCCCUCUCCAG |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-6787-3p | MIMAT0027475 |
Stem-loop ID | hsa-mir-6787 |
Stem-loop Accession # | MI0022632 |
Stem-loop Sequence | UCGGCUGGCGGGGGUAGAGCUGGCUGCAGGCCCGGCCCCUCUCAGCUGCUGCCCUCUCCAG |
Chromosome Location | Chr. 17 - 82236668 - 82236728 [+] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0027475
miRBase ID: hsa-miR-6787-3p Mature miRNA Sequence: UCUCAGCUGCUGCCCUCUCCAG Chromosome Location: Chr. 17 - 82236668 - 82236728 [+] on Build GRCh38 |