Assay Details
Amplicon Length: | 83 |
Chromosome Location: | Chr. 17 - 82236491 - 82236491 [+] on 82236491 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-6787-3p | MIMAT0027475 |
Human | hsa-miR-6787-5p | MIMAT0027474 |
Stem-loop Details
Human