Assay Details
Gene Family ID: | MIPF0000130, mir-506 |
Mature miRNA Sequence: | AAUGUGUAGCAAAAGACAGA |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-511-3p | MIMAT0026606 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-511-3p | ||
---|---|---|---|
miRBase Accession Number: | MI0003127 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.10: 17845107 - 17845193 [+] on Build GRCh38 | ||
Mature miRNA Sequence: | AAUGUGUAGCAAAAGACAGA | ||
Species: | Human | ||
Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0000130, mir-506 |
Mature miRNA Sequence: | AAUGUGUAGCAAAAGACAGA |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-511-3p | MIMAT0026606 |
Stem-loop ID | hsa-mir-511 |
Stem-loop Accession # | MI0003127 |
Stem-loop Sequence | CAAUAGACACCCAUCGUGUCUUUUGCUCUGCAGUCAGUAAAUAUUUUUUUGUGAAUGUGUAGCAAAAGACAGAAUGGUGGUCCAUUG |
Chromosome Location | Chr. 10 - 17845107 - 17845193 [+] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0026606
miRBase ID: hsa-miR-511-3p Mature miRNA Sequence: AAUGUGUAGCAAAAGACAGA Chromosome Location: Chr. 10 - 17845107 - 17845193 [+] on Build GRCh38 |