Assay Details
Amplicon Length: | 61 |
Chromosome Location: | Chr. 10 - 17844703 - 17844703 [+] on 17844703 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-511-3p | MIMAT0026606 |
Human | hsa-miR-511-5p | MIMAT0002808 |
Stem-loop Details
Human