Assay Details
Gene Family ID: | MIPF0001584, mir-3934 |
Mature miRNA Sequence: | UGCUCAGGUUGCACAGCUGGGA |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-3934-3p | MIMAT0022975 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-3934-3p | ||
---|---|---|---|
miRBase Accession Number: | MI0016590 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.6: 33698128 - 33698234 [+] on Build GRCh38 | ||
Mature miRNA Sequence: | UGCUCAGGUUGCACAGCUGGGA | ||
Species: | Human | ||
Product Type: | Ambion® Pre-miR™ miRNA Precursor |
Gene Family ID: | MIPF0001584, mir-3934 |
Mature miRNA Sequence: | UGCUCAGGUUGCACAGCUGGGA |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-3934-3p | MIMAT0022975 |
Stem-loop ID | hsa-mir-3934 |
Stem-loop Accession # | MI0016590 |
Stem-loop Sequence | CACAGCCCUUCCUGUCCCCAGUUUUCAGGUGUGGAAACUGAGGCAGGAGGCAGUGAAGUAACUUGCUCAGGUUGCACAGCUGGGAAGUGGAGCAGGGAUUUGAAUCC |
Chromosome Location | Chr. 6 - 33698128 - 33698234 [+] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0022975
miRBase ID: hsa-miR-3934-3p Mature miRNA Sequence: UGCUCAGGUUGCACAGCUGGGA Chromosome Location: Chr. 6 - 33698128 - 33698234 [+] on Build GRCh38 |