Assay Details
Amplicon Length: | 84 |
Chromosome Location: | Chr. 6 - 33697981 - 33697981 [-] on 33697981 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-3934-3p | MIMAT0022975 |
Human | hsa-miR-3934-5p | MIMAT0018349 |
Stem-loop Details
Human