Assay Details
Mature miRNA Sequence: | CUUGAGUCGUGCCUUUCUGAAUG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-8081 | MIMAT0031008 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-8081 | ||
---|---|---|---|
miRBase Accession Number: | MI0025917 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.9: 106600928 - 106601022 [+] on Build GRCh38 | ||
Mature miRNA Sequence: | CUUGAGUCGUGCCUUUCUGAAUG | ||
Species: | Human | ||
Product Type: | mirVana® miRNA inhibitor |
Mature miRNA Sequence: | CUUGAGUCGUGCCUUUCUGAAUG |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-8081 | MIMAT0031008 |
Stem-loop ID | hsa-mir-8081 |
Stem-loop Accession # | MI0025917 |
Stem-loop Sequence | UUAUUGUCUUUAUUUUCUGAGUCUUGCCUCUAUGGCCCCCCUCCUCUCUGAAAGCAAGGCCACUUGAGUCGUGCCUUUCUGAAUGAAGUCAAUGU |
Chromosome Location | Chr. 9 - 106600928 - 106601022 [+] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0031008
miRBase ID: hsa-miR-8081 Mature miRNA Sequence: CUUGAGUCGUGCCUUUCUGAAUG Chromosome Location: Chr. 9 - 106600928 - 106601022 [+] on Build GRCh38 |