Assay Details
Amplicon Length: | 85 |
Chromosome Location: | Chr. 9 - 106600745 - 106600745 [-] on 106600745 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-8081 | MIMAT0031008 |
Stem-loop Details
Human