Assay Details
Mature miRNA Sequence: | GCCCGAGAGGAUCCGUCCCUGC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4740-3p | MIMAT0019870 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-4740-3p | ||
---|---|---|---|
miRBase Accession Number: | MI0017378 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.17: 81400716 - 81400778 [-] on Build GRCh38 | ||
Mature miRNA Sequence: | GCCCGAGAGGAUCCGUCCCUGC | ||
Species: | Human | ||
Product Type: | mirVana® miRNA inhibitor |
Mature miRNA Sequence: | GCCCGAGAGGAUCCGUCCCUGC |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4740-3p | MIMAT0019870 |
Stem-loop ID | hsa-mir-4740 |
Stem-loop Accession # | MI0017378 |
Stem-loop Sequence | GCCAAGGACUGAUCCUCUCGGGCAGGGAGUCAGAGGGGACCGCCCGAGAGGAUCCGUCCCUGC |
Chromosome Location | Chr. 17 - 81400716 - 81400778 [-] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0019870
miRBase ID: hsa-miR-4740-3p Mature miRNA Sequence: GCCCGAGAGGAUCCGUCCCUGC Chromosome Location: Chr. 17 - 81400716 - 81400778 [-] on Build GRCh38 |