Assay Details
Amplicon Length: | 95 |
Chromosome Location: | Chr. 17 - 81400965 - 81400965 [+] on 81400965 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4740-3p | MIMAT0019870 |
Human | hsa-miR-4740-5p | MIMAT0019869 |
Stem-loop Details
Human