Assay Details
Amplicon Length: | 74 |
Chromosome Location: | Chr. 9 - 35710891 - 35710891 [-] on 35710891 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-6852-3p | MIMAT0027605 |
Human | hsa-miR-6852-5p | MIMAT0027604 |
Stem-loop Details
Human