Assay Details
Mature miRNA Sequence: | UGUCCUCUGUUCCUCAG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-6852-3p | MIMAT0027605 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-6852-3p | ||
---|---|---|---|
miRBase Accession Number: | MI0022698 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.9: 35710676 - 35710741 [-] on Build GRCh38 | ||
Mature miRNA Sequence: | UGUCCUCUGUUCCUCAG | ||
Species: | Human | ||
Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Mature miRNA Sequence: | UGUCCUCUGUUCCUCAG |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-6852-3p | MIMAT0027605 |
Stem-loop ID | hsa-mir-6852 |
Stem-loop Accession # | MI0022698 |
Stem-loop Sequence | UGCUGCCCUGGGGUUCUGAGGACAUGCUCUGACUCCCCUGAUGUCCUCUGUUCCUCAGGUGCUGGG |
Chromosome Location | Chr. 9 - 35710676 - 35710741 [-] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0027605
miRBase ID: hsa-miR-6852-3p Mature miRNA Sequence: UGUCCUCUGUUCCUCAG Chromosome Location: Chr. 9 - 35710676 - 35710741 [-] on Build GRCh38 |