Assay Details
Amplicon Length: | 115 |
Chromosome Location: | Chr. 12 - 48132620 - 48132620 [+] on 48132620 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-6505-3p | MIMAT0025467 |
Human | hsa-miR-6505-5p | MIMAT0025466 |
Stem-loop Details
Human