Assay Details
Gene Family ID: | MIPF0001805, mir-6505 |
Mature miRNA Sequence: | UGACUUCUACCUCUUCCAAAG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-6505-3p | MIMAT0025467 |
Stem-loop Details
Human
Test
Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | hsa-miR-6505-3p | ||
---|---|---|---|
Stem-loop Accession Number: | MI0022217 | ||
miRBase Version: | v22.1 | ||
Chromosome Location: | Chr.12: 48132797 - 48132867 [+] on Build GRCh38 | ||
Mature miRNA Sequence: | UGACUUCUACCUCUUCCAAAG | ||
Species: | Human | ||
Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0001805, mir-6505 |
Mature miRNA Sequence: | UGACUUCUACCUCUUCCAAAG |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-6505-3p | MIMAT0025467 |
Stem-loop ID | hsa-mir-6505 |
Stem-loop Accession # | MI0022217 |
Stem-loop Sequence | GCAUUGGAAUAGGGGAUAUCUCAGCAUGUUGAGCCCUGUCUCUGGGGAGCUGACUUCUACCUCUUCCAAAG |
Chromosome Location | Chr. 12 - 48132797 - 48132867 [+] on Build GRCh38 |
Mature MicroRNA |
miRBase Accession #: MIMAT0025467
miRBase ID: hsa-miR-6505-3p Mature miRNA Sequence: UGACUUCUACCUCUUCCAAAG Chromosome Location: Chr. 12 - 48132797 - 48132867 [+] on Build GRCh38 |